missing translation for 'onlineSavingsMsg'
Learn More

CHD3 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30500338 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500338 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500338 Supplier Novus Biologicals Supplier No. NBP337987FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHD3 Polyclonal antibody specifically detects CHD3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHD3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase CHD3, CHD-3, chromodomain helicase DNA binding protein 3, chromodomain-helicase-DNA-binding protein 3, EC 3.6.1, EC 3.6.4.12, hZFH, Mi-2 autoantigen 240 kDa protein, Mi-2a, Mi2-ALPHA, ZFH, Zinc finger helicase, zinc-finger helicase (Snf2-like)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1900-2000 of human CHD3 (NP_001005273.1).,, Sequence:, ERSILSRLASKGTEPHPTPAYPPGPYATPPGYGAAFSAAPVGALAAAGANYSQMPAGSFITAATNGPPVLVKKEKEMVGALVSDGLDRKEPRAGEVICIDD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune Dysfunction, Immune System Diseases, Immunology, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 1107
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.