Learn More
CHCHD3 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803432S
Encompass_Preferred
 View more versions of this product
Product Code. 17572493
  • 242.00 € / 30µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CHCHD3 Monoclonal antibody specifically detects CHCHD3 in Human samples. It is validated for Western Blot
Specifications
Specifications
CHCHD3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P63272
Mouse
Affinity chromatography
RUO
6827
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2G8
Unconjugated
SUPT4H1
0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; coiled-coil-helix-coiled-coil-helix domain containing 3; Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; Mic19; MICOS complex subunit MIC19; MINOS3; mitochondrial inner membrane organizing system 3; PPP1R22; protein phosphatase 1, regulatory subunit 22
Full length human recombinant protein of human SUPT4H1 produced in E.coli.
30 μL