Learn More
CHAC1 Monoclonal Antibody (OTI4B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507048
Description
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.Specifications
| CHAC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BUX1 | |
| CHAC1 | |
| Full length human recombit protein of human CHAC1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4B3 | |
| Unconjugated | |
| CHAC1 | |
| ChaC glutathione specific gamma-glutamylcyclotransferase 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79094 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.