missing translation for 'onlineSavingsMsg'
Learn More

CHAC1 Monoclonal Antibody (OTI1E2)
GREENER_CHOICE

Product Code. 15985904
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15985904 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15985904 Supplier Invitrogen Supplier No. MA526311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHAC1 Monoclonal Antibody (OTI1E2)

CHAC1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CHAC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHAC1
Gene Accession No. Q9BUX1
Gene Alias 1810008K03Rik; Blocks Notch protein; botch; cation transport regulator homolog 1; cation transport regulator-like protein 1; ChaC glutathione specific gamma-glutamylcyclotransferase 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Chac1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; glutathione-specific gamma-glutamylcyclotransferase 1; Neuroprotective protein 7; Npg7; RGD1307153
Gene Symbols CHAC1
Host Species Mouse
Immunogen Full length human recombinant protein of CHAC1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79094
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.