Learn More
Tocris Bioscience™ CGP 71683 hydrochloride
GREENER_CHOICE
Product Code. 18005412
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18005412
Brand: Tocris Bioscience™ 2199/50
Please to purchase this item. Need a web account? Register with us today!
Highly selective and potent non-peptide NPY Y5 receptor antagonist
CGP 71683 hydrochloride is an extremely selective, non-peptide NPY Y5 receptor antagonist. Displays > 1000-fold selectivity over Y1, Y2 and Y4 receptors; IC50 values are 1.4, 2765, 7187 and 5637 nM at cloned rat Y5, Y1, Y2 and Y4 receptors respectively. Potently inhibits NPY-induced food intake following i.p. administration in diabetic, free-feeding and fasted rats.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mg
Product Type CGP 71683 hydrochloride
Purity >99%
Molecular Weight (g/mol) 512.07
Inhibitors CGP 71683 hydrochloride