missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CFUrine Kit™
Description
CFUrine quality control kits are used for determination of urine colony counts for dipslides, tube screening methods and calibrated loops.
- Film of preserved microorganisms on the inside of the cap on plastic vial
- Ready for rehydration
Specifications
Specifications
| ATCC® | 25922TM* |
| Description | CFUrine Kit™ Escherichia coli |
| Format | Vials |
| Product Type | Quality Control Kit |
| Target Organism Class | Organism Sets |
| Strain Designations | FDA strain Seattle 1946; DSM 1103; NCIB 12210 |
| Species | Escherichia species |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?