Learn More
CFTR corrector 2, MedChemExpress
Product Code. 16455916
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16455916
Brand: Medchem Express HY125381
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CFTR corrector 2 is a cystic fibrosis transmembrane conductance corrector (CFTR), extracted from patent US20140274933.
CFTR corrector 2 is a cystic fibrosis transmembrane conductance corrector (CFTR), extracted from patent US20140274933.
Specifications
Chemical Name or Material CFTR corrector 2
CAS 1628416-28-3
Color White
Purity Grade Notes Research
Molecular Formula C27H23FN4O4
Quantity 100 mg
Solubility Information DMSO : 62.5 mg/mL (128.47 mM; Need ultrasonic)
SMILES O=C(N(C)C1=CC=C2N=C(C)OC2=C1)CN(N=C(C3=CC=CC(F)=C3)C4=C5C=C(OCC)C=C4)C5=O
Molecular Weight (g/mol) 486.49
Formula Weight 486.49
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation