Learn More
Cevipabulin, MedChemExpress
Product Code. 16466268
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16466268
Brand: Medchem Express HY14949
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Cevipabulin (TTI-237) is an oral, microtubule-active antitumor compound and inhibits the binding of [3H] vinblastine to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell line.
Cevipabulin (TTI-237) is an oral, microtubule-active antitumor compound and inhibits the binding of [3H] vinblastine to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell line.
Specifications
Chemical Name or Material Cevipabulin
CAS 849550-05-6
Color White
Purity Grade Notes Research
Molecular Formula C18H18ClF5N6O
Quantity 10 mg
Synonym TTI-237
Solubility Information DMSO : 16.67 mg/mL (35.86 mM; Need ultrasonic)
SMILES C[C@@]([H])(C(F)(F)F)NC1=C(C2=C(F)C=C(OCCCNC)C=C2F)C(Cl)=NC3=NC=NN13
Molecular Weight (g/mol) 464.82
Formula Weight 464.82
Percent Purity 95.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death