missing translation for 'onlineSavingsMsg'
Learn More

Cesium chromate, 99+%, Thermo Scientific™

Product Code. p-7027112
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11495647

Brand: Thermo Scientific Alfa Aesar B23619.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-78-9
Molecular Formula CrCs2O4
Molecular Weight (g/mol) 381.80
MDL Number MFCD00016050
InChI Key BROHICCPQMHYFY-UHFFFAOYSA-N
Synonym cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
PubChem CID 61613
IUPAC Name dicesium;dioxido(dioxo)chromium
SMILES [Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 10 g
UN Number UN3087
Formula Weight 381.81
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium chromate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.