missing translation for 'onlineSavingsMsg'
Learn More

Ceritinib dihydrochloride, MedChemExpress

Product Code. 16436728
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436728

Brand: Medchem Express HY15656A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ceritinib dihydrochloride (LDK378 dihydrochloride) is a selective, orally bioavailable and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib dihydrochloride (LDK378 dihydrochloride) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib dihydrochloride (LDK378 dihydrochloride) shows great antitumor potency.

Ceritinib dihydrochloride (LDK378 dihydrochloride) is a selective, orally bioavailable and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib dihydrochloride (LDK378 dihydrochloride) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib dihydrochloride (LDK378 dihydrochloride) shows great antitumor potency.

Specifications

Chemical Name or Material Ceritinib dihydrochloride
CAS 1380575-43-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H38Cl3N5O3S
Quantity 10 mg
Synonym LDK378 dihydrochloride
Solubility Information DMSO : 100 mg/mL (158.46 mM; Need ultrasonic) ∣H2O : 10 mg/mL (15.85 mM; Need ultrasonic)
SMILES CC(C)OC1=CC(C2CCNCC2)=C(C)C=C1NC3=NC=C(Cl)C(NC4=CC=CC=C4S(=O)(C(C)C)=O)=N3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 631.06
Formula Weight 631.06
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.