missing translation for 'onlineSavingsMsg'
Learn More

Cerebellin-4 Antibody [DyLight 488], Novus Biologicals™

Product Code. 30734432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734432 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734432 Supplier Novus Biologicals Supplier No. AF6740K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cerebellin-4 Polyclonal antibody specifically detects Cerebellin-4 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cerebellin-4
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CBLNL1, cerebellin 4 precursor, cerebellin precursor-like 1, cerebellin-4, Cerebellin-like glycoprotein 1, dJ885A10.1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Cerebellin-4, Gln28-Leu201, Accession # Q9NTU7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140689
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.