Learn More
Cephalotaxine, TRC
Specifications
Specifications
| Chemical Name or Material | Cephalotaxine |
| CAS | 24316-19-6 |
| Molecular Formula | C18 H21 N O4 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C18H21NO4/c1-21-15-9-18-4-2-5-19(18)6-3-11-7-13-14(23-10-22-13)8-12(11)16(18)17(15)20/h7-9,16-17,20H,2-6,10H2,1H3/t16-,17-,18+/m1/s1 |
| SMILES | COC1=C[C@]23CCCN2CCc4cc5OCOc5cc4[C@@H]3[C@@H]1O |
| IUPAC Name | (2S,3S,6R)-4-methoxy-16,18-dioxa-10-azapentacyclo[11.7.0.02,6.06,10.015,19]icosa-1(20),4,13,15(19)-tetraen-3-ol |
| Molecular Weight (g/mol) | 315.36 |
| Formula Weight | 315.1471 |