Learn More
Cephalotaxine, MedChemExpress
Description
Cephalotaxlen ((-)-Cephalotaxine) is an alkaloid that can be isolated from Cephalotaxus drupacea, with antileukemic and antiviral activities. Cephalotaxlen has anti-ZIKV (Zika virus) activity.
Specifications
Specifications
| Chemical Name or Material | Cephalotaxine |
| Physical Form | Powder |
| CAS | 24316-19-6 |
| Color | Light Nude |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H21NO4 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | (-)-Cephalotaxine ZINC19795976 |
| Solubility Information | DMSO : 25 mg/mL (79.27 mM; Need ultrasonic) |
| Show More |