Learn More
Invitrogen™ CEP63 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113199
This item is not returnable.
View return policy
Description
CEP63 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
Specifications
| CEP63 | |
| Polyclonal | |
| Unconjugated | |
| CEP63 | |
| 4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 28135, 80254 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3UPP8, Q96MT8 | |
| CEP63 | |
| Recombinant Human Centrosomal protein of 63 kDa protein (1-495AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |