Learn More
MedChemExpress CEP dipeptide 1
Specifications
Specifications
| Purity | 98.0% |
| CAS | 816432-15-2 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 381.42 g/mol |
| Product Type | Peptides |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 10 mg/mL (26.22 mM∣Need ultrasonic) |