missing translation for 'onlineSavingsMsg'
Learn More

CEP-28122 mesylate salt, MedChemExpress

Product Code. p-200102001
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499927

Brand: Medchem Express HY18030A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CEP-28122 mesylate salt, a diaminopyrimidine derivative, is a potent, selective, and orally bioavailable ALK inhibitor, with an IC50 value of 1.9 nM for recombinant ALK kinase activity. CEP-28122 has antitumor activity in experimental models of ALK-positive human cancers. CEP-28122 mesylate salt has good pharmacodynamic and pharmacokinetic activity.

CEP-28122 mesylate salt, a diaminopyrimidine derivative, is a potent, selective, and orally bioavailable ALK inhibitor, with an IC50 value of 1.9 nM for recombinant ALK kinase activity. CEP-28122 has antitumor activity in experimental models of ALK-positive human cancers. CEP-28122 mesylate salt has good pharmacodynamic and pharmacokinetic activity.

Specifications

Chemical Name or Material CEP-28122 mesylate salt
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H39ClN6O6S
Quantity 5 mg
Solubility Information DMSO : ≥ 6.4 mg/mL (10.08 mM)
SMILES O=C([C@H]1[C@](C2)([H])C=C[C@]2([H])[C@H]1NC3=NC(NC4=CC=C5C(CC[C@@H](N6CCOCC6)CC5)=C4OC)=NC=C3Cl)N.O=S(C)(O)=O
Molecular Weight (g/mol) 635.17
Formula Weight 635.17
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.