Learn More
Invitrogen™ CellVue™ Maroon Cell Labeling Kit
Product Code. 11540637
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 11540637
Brand: Invitrogen™ 88087016
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Far-red fluorescent reagent used to label the cell membrane in order to identify and track labeled cells. Cell labeling is rapid and stable and can be combined with fluorescently labeled antibodies and other markers of cellular function for flow cytometric analysis and fluorescent microscopy.
  • The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
  • CellVue™ Maroon is optimally excited at 647nm and has a peak emission of 667nm
  • It can be detected equally in filter sets designed for APC, Alexa Fluor™ 700, and APC-eFluor™ 780 or similar fluorochromes – using antibodies conjugated to these fluorochromes in combination with CellVue™ Maroon is not recommended
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 kit
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Label Type eFluor
Product Type Cell Labeling
For Research Use Only. Not for use in diagnostic procedures.