Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow are recommended for detection of yeasts and molds, particle count, and microscopy.
Brand: Sartorius Stedim Biotech™ 130H6--50----ACR
Additional Details : Weight : 1.36900kg ??product.attributeSet.Transport?? : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Sterile, individually packaged filters
- Save preparatory time
- Perfect grid printing
- Filter colors allow the best contrast to the colonies or particles
- Outstanding microorganism recovery rate
- High flow membranes
- Sterilized by gamma irradiation
- Compatible with aqueous solutions (pH 4-8), hydrocarbons and several other organic solvents
Specifications
Gray/White Grid | |
0.45 μm | |
130 | |
115 to 145 μm |
Cellulose Nitrate (Cellulose Mixed Ester) | |
Sterile | |
1000/pk. | |
50 mm |