Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Shop All Macherey-Nagel Products
Click to view available options
Height (Metric):
94 mm
205 mm
Unit Size:
Pack of 25
Specifications
Specifications
| Diameter (Metric) Inside | 33 mm |
| Material | Cellulose |
| For Use With (Application) | Analysis of soil contaminants; determination of binder content in asphalt; fat determination in food and feed |
| Grade | 645 |
| For Use With (Equipment) | Soxhlet extractors or automised extractors |
| Height (Metric) | 94 mm |