missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cellulose Dialysis Tubing Roll
Click to view available options
Diameter (Metric):
14.3 mm
Unit Size:
Pack of 30
Specifications
Specifications
| Product Type | Dialysis Tubing Roll |
| MWCO | 12 to 14 kDa |
| Material | Cellulose |
| Diameter (Metric) | 14.3 mm |
| Length (Metric) | 30 m |
| Color | Transparent |
| Quantity | 30 Pack |
| For Use With (Application) | For viscose process |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction