Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Non-cytotoxic
- Compatible with automated systems
- Flexible working volume
- Rounded square well profile
- Stackable
- Alphanumeric well cod
Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Packaging | 10/Pk., 40/Cs. |
| Color | Black, Clear Bottom |
| Product Type | Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 15 to 110 μL |
| Show More |