Learn More
Ion Torrent™ Celleste™ Image Analysis Software, network seat, one additional seat
Product Code. 16646484
Click to view available options
Quantity:
1 license
missing translation for 'unitSize'
Each
Product Code. 16646484
missing translation for 'mfr': Ion Torrent™ AMEP4869
Please to purchase this item. Need a web account? Register with us today!
Celleste 5.0 Image Analysis Software is a full-feature package for a range of biological applications with segmentation and classification tools that easily transform images into quantitative data in a streamlined workflow. The Celleste 5.0 Image Analysis Network Seat is designed for environments where the use of multiple copies is desired. It requires an active Celleste 5.0 Network License Server License. Optional modules for 2D and 3D deconvolution, 3D visualization and analysis allow you to customize the capabilities according to your needs.
Celleste 5.0 Image Analysis Software has features designed for a range of biological applications, such as image adjustment and processing with manual and automatic measurements over multiple channels, and segmentation and classification tools that help you transform images into quantitative data in a streamlined and customizable workflow. Optional modules for deconvolution, 3D visualization, and 3D analysis allow you to customize the capabilities according to your needs. Coupled with the powerful image acquisition capabilities of our EVOS microscopes, Celleste software allows you to seamlessly capture, process, measure, analyze, and share images and data.
This Network Seat is designed for environments where the use of multiple copies is desired. It requires an active Celleste 5.0 Network Server License.
Celleste 5.0 Image Analysis Software offers you these important advantages:
• Streamlined workflow tools that make it easy to visualize and edit images
• Comprehensive image processing and enhancement functions with optional modules to meet your specific needs
• Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images
• A new interface for streamlined multi-well plate navigation and well selection
• A wide range of functions to create reports and share images and data
Process and analyze
After image acquisition, immediately apply a suite of image processing and enhancement functions with a few clicks of a mouse. Use auto-alignment for overlays and remove background to reveal critical details with a wide range of filters. Once satisfied with the results from one image, apply the same steps to a set of time-lapse images, scans, or results from different treatments. When imaging cells in multi-well plates, use the new interface to select only the wells you wish to analyze and automatically process them as a set.
Measure and quantify
Easily measure and analyze your images with a variety of measurement tools, such as those for distance, region, angles, and area. The ability to identify an object in time-lapse experiments can be used to track cell movement or migration. This feature is particularly powerful in combination with the EVOS Onstage Incubator, which allows continuous monitoring of cells under controlled environmental conditions.
Count and classify
Using a number of manual and automatic measurement tools, easily count and characterize cells and subcellular objects based on area, length, shape, and light intensity. Flexible segmentation tools allow you to label images based on color or intensity level. A new ROI functionality allows you to count objects and maintain measurements by region (e.g., a parent-child application) and also to count by ROI. After object counting, sort and display counted images based on size and other parameters.
Report and share
Upon completing the image analysis, a suite of annotation and reporting tools allow you to create presentation-ready image and data reports with a few clicks and to share them with others in PDF, PowerPoint, and Excel format.
Celleste 5.0 software is a powerful image analysis solution that helps make your imaging research job easier and increase your productivity by seamlessly providing images and accurate data. With the proven usability of EVOS microscopes, you now have the tools to make it easy to capture, process, measure, analyze, and share your important images and data more effectively than ever.
Compliance
RUO
[1] http://www.thermofisher.com/Celleste[2] Celleste Image Analysis Network License [AMEP4868]Explore the entire EVOS line of imaging systems, accessories, and offers [3] [www.thermofisher.com/EVOS][4] EVOS Onstage Incubator [www.thermofisher.com/AMC1000]In-the boxOne Celleste 5.0 Image Analysis Network SeatFor Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Celleste
For Use With (Equipment) EVOS M5000, EVOSM7000 and Other Imaging Systems
Quantity 1 license