missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLCOAT™ Poly-L-lysine Coated Cell Culture Dish

Product Code. 11759442
Click to view available options
Diameter (Metric):
60 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11759442

Brand: Greiner Bio-One 628930

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cultivation in serum-free and serum reduced medium. Greiner Bio-One CELLCOAT™ Poly-L-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, serum-free or -reduced cell culture and general improvement o

  • Source: Synthetic
  • Cultivation of neuronal cells, transfected cells and other fastidious cell lines
  • Cultivation in serum-free and serumreduced medium
  • Improvement of cell attachment
  • Shelf life at room temperature:
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 28 mL
Color Clear
Culture Area 21 sq. cm
Material Polystyrene
Shape Round
Sterility Non-sterile
Surface Treatment Poly-L-Lysine
Type Cell Culture Dish
Volume (Metric) Working 6 to 7 mL
Includes Lid
Height (Metric) 15 mm
Diameter (Metric) 60 mm
Show More Show Less

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.