missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLCOAT™ 96-well, Poly-L Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Product Code. 13092213
Click to view available options
Color:
Clear
Unit Size:
Pack of 20
This item is not returnable. View return policy

Product Code. 13092213

Brand: Greiner Bio-One 655930

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Clear; Sterility:Non-sterile; Lid:With Lid

Alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Culture Area 34 mm2
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Non-sterile
Surface Treatment Poly-L-Lysine
Well Shape Flat Bottom (F-well)
For Use With (Application) Cell Culture
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 20
No. per Pack 5
Quantity 20/Cs.
Temperature Range (Metric) Room Temperature
Well Design Chimney
Dimensions (L x W x H) 128 x 85 x 19 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.