missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CellAdhere™ Laminin-521

Product Code. 15844755
Encompass_Preferred
Click to view available options
Quantity:
10 x 100 μg
100 μg
Unit Size:
1000µg
100µg
This item is not returnable. View return policy

Product Code. 15844755

Brand: STEMCELL Technologies 77003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Matrix for maintenance of human ES and iPS cells in combination with mTeSR™1, TeSR™-E8™, or TeSR™2

  • Support the growth and differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under feeder-free conditions with this defined and xeno-free cell culture matrix
  • For consistent cell populations and reproducible results in downstream applications, use CellAdhere™ Laminin-521 with TeSR™ maintenance media to provide a defined culture substrate for cell maintenance
  • Laminin 521 is expressed and secreted by human pluripotent stem cells (hPSCs) in the inner cell mass of the embryo and therefore creates a biologically relevant hPSC culture environment in vitro
  • Compared to other matrices, CellAdhere™ Laminin-521 increases single-cell attachment and survival and does not require the addition of apoptotic inhibitors during plating
  • Note that single-cell passaging of human ES and iPS cells can result in selective pressure and lead to genetic aberrations
  • If passaging as single cells, we recommend checking the karyotype frequently

Specifications

Cell Type Pluripotent Stem Cells
Quantity 100 μg
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.