Learn More
Celestolide-d6 (major), TRC
Specifications
Specifications
| Chemical Name or Material | Celestolide-d6 |
| CAS | 2733264-86-1 |
| Molecular Formula | C17 D6 H18 O |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C17H24O/c1-11(18)14-9-12(16(2,3)4)10-15-13(14)7-8-17(15,5)6/h9-10H,7-8H2,1-6H3/i5D3,6D3 |
| SMILES | [2H]C([2H])([2H])C1(CCc2c(cc(cc12)C(C)(C)C)C(=O)C)C([2H])([2H])[2H] |
| IUPAC Name | 1-[6-tert-butyl-1,1-bis(trideuteriomethyl)indan-4-yl]ethanone |
| Molecular Weight (g/mol) | 250.41 |
| Formula Weight | 250.2204 |