Learn More
Cefmenoxime hydrochloride, MedChemExpress
Product Code. 16482216
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16482216
Brand: Medchem Express HYB0875
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Cefmenoxime (SCE-1365) hydrochloride is a new semisynthetic cephalosporin antibiotic. Cefmenoxime has antibacterial activity against a wide variety of gram-positive and gram-negative bacteria.
Cefmenoxime (SCE-1365) hydrochloride is a new semisynthetic cephalosporin antibiotic. Cefmenoxime has antibacterial activity against a wide variety of gram-positive and gram-negative bacteria.
Specifications
Chemical Name or Material Cefmenoxime hydrochloride
Physical Form Solid
CAS 75738-58-8
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C16H17N9O5S3.1/2 HCl
For Use With (Application) COVID-19-immunoregulation
Synonym SCE-1365 hydrochloride
Solubility Information DMSO : 65 mg/mL (122.69 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(C(N12)=C(CSC3=NN=NN3C)CS[C@]2([H])[C@H](NC(/C(C4=CSC(N)=N4)=N\OC)=O)C1=O)O.[0.5HCl]
Molecular Weight (g/mol) 529.79
Formula Weight 529.79
Percent Purity 98.11%
Grade Research