missing translation for 'onlineSavingsMsg'
Learn More

Cefetamet pivoxil hydrochloride, MedChemExpress

Product Code. p-200099332
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16438826

Brand: Medchem Express HYB1894A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cefetamet pivoxil hydrochloride is an oral third generation cephalosporin antibiotic.

Cefetamet pivoxil hydrochloride is an oral third generation cephalosporin antibiotic.

Specifications

Chemical Name or Material Cefetamet pivoxil hydrochloride
Physical Form Solid
CAS 111696-23-2
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H26ClN5O7S2
For Use With (Application) COVID-19-immunoregulation
Synonym Ro 15-8075
Solubility Information DMSO : ≥ 100 mg/mL (182.47 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C(N12)=C(C)CS[C@]2([H])[C@H](NC(/C(C3=CSC(N)=N3)=N\OC)=O)C1=O)OCOC(C(C)(C)C)=O.[H]Cl
Molecular Weight (g/mol) 548.03
Formula Weight 548.03
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.