missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ceapin A7

Description
Ceapin A7 is a selective inhibitor of ATF6α (IC50=0.59 μM). The compound selectively inhibits ATF6α over ATF6β and other branches of the unfolded protein response (UPR). Ceapin A7 inhibits ER stress-induced upregulation of BiP in vitro. Ceapin A7 inhibits cleavage and functional activation of endogenous ATF6 in response to ER stress in vitro.
Specifications
Specifications
| Chemical Name or Material | N-[1-[[2,4-bis(Trifluoromethyl)phenyl]methyl]-1 H-pyrazol-4-yl]-5-(2-furanyl)-3-isoxazolecarboxamide |
| CAS | 2323027-38-7 |
| Quantity | 10 mg |
| Target | Other ER Stress/UPR Activators |
| Molecular Formula | C20H12F6N4O3 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?