missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM6 Monoclonal Antibody (A1E2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537801
This item is not returnable.
View return policy
Description
Positive controls: JAR cell lysate, SK-Br-3 cell lysate, A549, HT-29, MCF-7, human lung tissue, human colon tissue, human colon carcinoma tissue, SW620.
CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
Specifications
| CEACAM6 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide, pH 7.4 | |
| P40199 | |
| CEACAM6 | |
| Recombinant protein within Human CEACAM6 aa 1-160. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| A1E2 | |
| Unconjugated | |
| CEACAM6 | |
| BGP; BGP-1; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66c; CD66d; CD66e; CEA; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; Cluster of Differentiation 66c; NCA; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM | |
| Mouse | |
| Protein A | |
| RUO | |
| 4680 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction