Learn More
CDKN1A Monoclonal Antibody (OTI4B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808276S
Description
CDKN1A Monoclonal antibody specifically detects CDKN1A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CDKN1A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Activating Fragment 1; CAP20; cdk inhibitor CIP1; cdk inhibitor CIP1 (p21); CDKI; CDK-interacting protein 1; CDK-interaction protein 1; Cdkn1; CDKN1A; CDNK1A; CIP1; Cip1); cyclin dependent kinase inhibitor 1A; cyclin-dependent kinase inhibitor 1; cyclin-dependent kinase inhibitor 1A; Cyclin-dependent kinase inhibitor 1A (p21; cyclin-dependent kinase inhibitor 1A (P21); cyclin-dependent kinase inhibitor 1A (p21, Cip1); cyclin-dependent kinase inhibitor 1A variant 1; cyclin-dependent kinase inhibitor 1A variant 2; DNA synthesis inhibitor; MDA6; MDA-6; melanoma differentiation associated protein 6; melanoma differentiation-associated protein; Melanoma differentiation-associated protein 6; OTTHUMP00000016298; p21; p21<WAF1/CIP1>; p21Cip1; p21Cip1/Waf1; p21WAF; PIC1; putative cyclin-dependent kinase inhibitor 1A (p21/WAF1/CIP1); SDI1; SLC12A9; UV96; WAF1; Wild type p53 activated fragment 1 (WAF1); wild-type p53-activated fragment 1 | |
| Full length human recombinant protein of human MGC12965 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B11 | |
| Unconjugated | |
| LOC100287896 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100287896 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.