missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK1/2/3, p Thr14 Antibody (SR1566), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22647-100ul
This item is not returnable.
View return policy
Description
CDK1/2/3 Monoclonal antibody specifically detects CDK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| CDK1/2/3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| A synthesized peptide derived from human Phospho-CDK1/2/3 [Thr14] (Uniprot #: P06493, P24941, Q00526) | |
| 100 μg | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1566 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction