Learn More
CDDO-Im, MedChemExpress
Product Code. 16499317
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16499317
Brand: Medchem Express HY15725
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CDDO-Im (RTA-403) is an activator of Nrf2 and PPAR, with Kis of 232 and 344 nM for PPARα and PPARγ.
CDDO-Im (RTA-403) is an activator of Nrf2 and PPAR, with Kis of 232 and 344 nM for PPARα and PPARγ.
Specifications
Chemical Name or Material CDDO-Im
CAS 443104-02-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C34H43N3O3
Quantity 5 mg
Synonym RTA-403 TP-235 CDDO-Imidazolide
Solubility Information DMSO : 50 mg/mL (92.30 mM; Need ultrasonic)
SMILES CC(C)([C@]1([H])CC[C@@]([C@@]2(CC[C@]3(CCC(C)(C[C@@]3([H])[C@]24[H])C)C(N5C=CN=C5)=O)C)6C)C(C(C#N)=C[C@]1(C)C6=CC4=O)=O
Molecular Weight (g/mol) 541.72
Formula Weight 541.72
Percent Purity 95.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death