missing translation for 'onlineSavingsMsg'
Learn More

CDDO-Im, MedChemExpress

Product Code. 16416778
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16416778

Brand: Medchem Express HY15725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CDDO-Im (RTA-403) is an activator of Nrf2 and PPAR, with Kis of 232 and 344 nM for PPARα and PPARγ.

CDDO-Im (RTA-403) is an activator of Nrf2 and PPAR, with Kis of 232 and 344 nM for PPARα and PPARγ.

Specifications

Chemical Name or Material CDDO-Im
CAS 443104-02-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C34H43N3O3
Quantity 10 mg
Synonym RTA-403 TP-235 CDDO-Imidazolide
Solubility Information DMSO : 50 mg/mL (92.30 mM; Need ultrasonic)
SMILES CC(C)([C@]1([H])CC[C@@]([C@@]2(CC[C@]3(CCC(C)(C[C@@]3([H])[C@]24[H])C)C(N5C=CN=C5)=O)C)6C)C(C(C#N)=C[C@]1(C)C6=CC4=O)=O
Molecular Weight (g/mol) 541.72
Formula Weight 541.72
Percent Purity 95.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.