missing translation for 'onlineSavingsMsg'
Learn More

CDD3506, MedChemExpress

Product Code. 16443218
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443218 10 mg 10mg
16474486 100 mg 100mg
16465947 5 mg 5mg
16439766 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16443218 Supplier Medchem Express Supplier No. HY100902
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CDD3506 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity.

CDD3506 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity.

Specifications

Chemical Name or Material CDD3506
CAS 197913-15-8
Color Light Nude
Purity Grade Notes Research
Molecular Formula C22H19N3
Quantity 10 mg
Solubility Information DMSO : 19 mg/mL (58.39 mM; Need ultrasonic)
SMILES NC1=CN(C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C=N1
Molecular Weight (g/mol) 325.41
Formula Weight 325.41
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.