Learn More
CDCA3 Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811696
Description
CDCA3 is a F-box-like protein which is required for entry into mitosis. It acts by participating to E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase.Specifications
| CDCA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99618 | |
| CDCA3 | |
| Human recombit protein fragment corresponding to amino acids 152-252 of human CDCA3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G7 | |
| Unconjugated | |
| CDCA3 | |
| GRCC8, TOME-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 83461 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.