missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18501AF700
This item is not returnable.
View return policy
Description
CDCA2 Polyclonal antibody specifically detects CDCA2 in Human samples. It is validated for Western Blot
Specifications
| CDCA2 | |
| Polyclonal | |
| Western Blot | |
| cell division cycle associated 2, cell division cycle-associated protein 2, FLJ25804, MGC129906, MGC129907, protein phosphatase 1 [gamma] (PPP1R3A) binding protein Repo-Man, Repo-ManRecruits PP1 onto mitotic chromatin at anaphase protein | |
| Recognizes region between residues Between between 200 and 250.. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 157313 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction