missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDC7/DBF4 Recombinant Human Protein

CDC7 is a homolog protein that is critical for the G1/S transition and is also essential for initiation of DNA replication as cell division occurs. Recombinant full-length human CDC7 and DBF4 proteins were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tags.

Brand:  Invitrogen™ PV6275

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15428933

  • 1678.00 € / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

CDC7 (cell division cycle 7) is expressed in many normal tissues, but the overexpression of CDC7 may be associated with neoplastic transformation for some tumors and transformed cell lines. CDC7/DBF4 kinase promotes S phase by alleviating an inhibitory activity in Mcm4 that evolved to integrate several protein kinase.

  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications

Specifications

Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
91.2/103.5kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Recombinant
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol
100μg
Human
GST Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions