missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD90.2 (Thy-1.2) Monoclonal Antibody (53-2.1), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Invitrogen™ M028T03Y04-A

 View more versions of this product

Product Code. 30056257

  • 350.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD90.2 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90.2 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90.2 is an 18 kDa plasma membrane antigen expressed by hematopoietic cells and neurons in all species studied, highly expressed in connective tissue, various fibroblast and stromal cell lines and lastly, by tumor endothelial cell lines. In the mouse, CD90.2 is expressed on all thymocytes and peripheral T lymphocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD90.2 (Thy-1.2)
Monoclonal
0.1 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01831
Thy1
Affinity chromatography
RUO
21838
4°C, store in dark, DO NOT FREEZE!
Liquid
IgG2a κ
Flow Cytometry
53-2.1
NovaFluor Yellow 660
Thy1
CD90; FLJ33325; T25; Thy 1.2; Thy1; Thy-1; Thy-1 antigen; thy-1 membrane glycoprotein; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta
Rat
100 μg
Primary
Mouse
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.