missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8/CD3 Mouse anti-Human, APC,FITC,PE,PerCP-Cy™,¢5.5, Clone: SK7 (also known as Leu-4),X39,X40,SK1, BD FastImmune™
Mouse Monoclonal Antibody
Brand: BD Biosciences 346047
This item is not returnable.
View return policy
Description
CD8/CD3 Monoclonal antibody specifically detects CD8/CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD8/CD3 | |
| Monoclonal | |
| APC/FITC/PE/PerCP-Cyanine5.5 | |
| 50 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ, IgG2a κ |
| Flow Cytometry | |
| SK7 (also known as Leu-4), X39, X40, SK1 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction