missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Antibody (CD8/7793R), Novus Biologicals™
Rabbit Monoclonal Antibody
291.00 € - 538.00 €
Specifications
| Antigen | CD8 |
|---|---|
| Clone | CD8/7793R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691144
|
Novus Biologicals
NBP3-23705-20ug |
20 μg |
291.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698834
|
Novus Biologicals
NBP3-23705-100ug |
100 μg |
538.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD8 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 925 | |
| IgG κ | |
| Protein A or G purified |
| CD8/7793R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
| Recombinant human full-length CD8 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title