missing translation for 'onlineSavingsMsg'
Learn More

CD79a Mouse anti-Human, PE, Clone: HM47, BD

Product Code. 15871489
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15871489 25 Tests Each
15838538 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15871489 Supplier BD Biosciences Supplier No. 561942

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The HM47 monoclonal antibody specifically binds to the cytoplasmic domain of CD79a. CD79a is a 47 kDa type 1 transmembrane glycoprotein present on B lymphocytes. CD79a is also referred to as mb-1, IGA and Ig-alpha (Ig α ). It is expressed on B cells at various stages of differentiation, from pre-B cell stage, probably before expression of cytoplasmic µ chain, to plasma cell stage, in which it is detected only in the cytoplasm. CD79a associates with CD79b to form part of the B-cell receptor complex. It has been suggested that CD79a may play a role in mediating the transport of IgM to the cell surface. This antibody has been found to crossreact with permeabilized A20 cells (mouse B cell line).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a
Applications Flow Cytometry
Classification Monoclonal
Clone HM47
Conjugate PE
Description CD79a, immunoglobulin-associated alpha ; CD79A; Ig-alpha; IGA; Igα; MB-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD79a Peptide
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.