missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63, Mouse anti-Human, CF-Blue, Clone: TEA3/18, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15362.100Reactions
This item is not returnable.
View return policy
Specifications
| CD63 | |
| Monoclonal | |
| CF-Blue | |
| In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
| LAMP-3/ME491/MLA1/OMA81H/TSPAN30 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| TEA3/18 | |
| Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
| CD63 molecule | |
| CD63 | |
| Cell preparation of human cytochrome B enriched cells. | |
| RUO | |
| 967 | |
| Store in the dark at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction