missing translation for 'onlineSavingsMsg'
Learn More

CD54 Mouse anti-Human, PE, Clone: HA58, BD

Mouse Monoclonal Antibody

100.00 € - 223.63 €

Specifications

Antigen CD54
Clone HA58
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15840649
View Documents
BD Biosciences
560971
25 Tests
ONLINE EXCLUSIVE PRICE
107.00 €
100.00 € / Each
Save 7.00 €
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15868228
View Documents
BD Biosciences
555511
100 Tests
ONLINE EXCLUSIVE PRICE
224.00 €
223.63 € / Each
Save 0.37 €
0.17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.

Specifications

Specifications

CD54
Flow Cytometry
PE
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
ICAM-1; intercellular adhesion molecule 1; BB2; P3.58
HA58
Monoclonal
Mouse
Human
IFN-γ-treated BM314 colonic carcinoma cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.