missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD5 Antibody (Magenta patent anti-CD5) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00 € - 920.00 €
Specifications
| Antigen | CD5 |
|---|---|
| Clone | Magenta patent anti-CD5 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044167
|
Novus Biologicals™
NBP3-28212-50ug |
50 μg |
213.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047448
|
Novus Biologicals™
NBP3-28212-1mg |
1 mg |
920.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044385
|
Novus Biologicals™
NBP3-28212-100ug |
100 μg |
338.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| CD5 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
| CD5 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Magenta patent anti-CD5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P06127 | |
| 921 | |
| IgG | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title