missing translation for 'onlineSavingsMsg'
Learn More

CD49f (Integrin α6) Rat, anti-Mouse, Pacific Blue, Clone: GoH3, STEMCELL Technologies

Product Code. 17108701
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17108701 100 Tests 100 tests
17118701 25 Tests 25 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17108701 Supplier Stemcell Technologies Supplier No. 60037PB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

  • The GoH3 antibody reacts with CD49f (integrin α6), an ∼150 kDa transmembrane glycoprotein that associates non-covalently with CD29 (integrin β1) or CD104 (integrin β4) to form the heterodimeric receptors VLA-6 and α6β4, which bind the extracellular matrix protein laminin
  • CD49f is a disulfide-linked dimer comprising an ∼120 kDa heavy chain and an ∼30 kDa membrane-bound light chain
  • Splice variants exist, which affect the cytoplasmic domain of the protein
  • CD49f is expressed on the surface of T cells, monocytes, platelets, placental trophoblasts, epithelial cells, and endothelial cells
  • It is involved in cell adhesion and regulating signaling pathways involved in a variety of processes, including the activation and proliferation of T cells, and the differentiation and maintenance of stem cell pluripotency
  • CD49f is considered the most important marker for selecting mouse mammary stem and progenitor cells
  • The GoH3 antibody reacts with an extracellular epitope on CD49f

Specifications

Antigen CD49f (Integrin α6)
Applications Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay
Classification Monoclonal
Clone GoH3
Conjugate Pacific Blue
Host Species Rat
Quantity 100 Tests
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Pig, Human
Isotype IgG2a κ
Cell Type Mammary Cells
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.