Learn More
CD47 Rat anti-Mouse, Functional Grade, Clone: miap301, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-0471-95
Description
The BMA12 monoclonal antibody reacts with mouse CD100, also known as Sema4D. Sema4D/CD100 is a 150kDa transmembrane protein expressed constitutively on T cells and is involved in the activation of B cells through its interaction with CD72. Interaction of these two proteins diminishes the negative signaling normally effected by CD72. CD100 is already expressed strongly on resting T cells and somewhat weakly on B cells, but activation upregulates expression on both cell types. Antigen expression has been reported in Spleen, thymus and l
CD47 Monoclonal antibody specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Neutralization
Specifications
| CD47 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| Q61735 | |
| Rat | |
| 10 mg | |
| Primary | |
| Mouse | |
| Antibody |
| Flow Cytometry, Functional Assay, Neutralization | |
| miap301 | |
| Functional Grade | |
| CD47 | |
| 9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
| Affinity chromatography | |
| RUO | |
| 16423 | |
| 4° C | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.