missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Rabbit, Clone: PTPRC/1783R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 5788-RBM23-P1
Additional Details : Weight : 0.01000kg
Specifications
CD45RB | |
Cell Surface and Cytoplasmic | |
PTPRC/1783R | |
Unconjugated | |
NP_002829 | |
Rabbit | |
180-220kDa | |
Protein A/G Purified | |
RUO | |
5788 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
PTPRC | |
Human B-lymphocytes | |
Ramos, U-698, or GA-10 cells. Tonsil | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |