Learn More
CD45 Rat, FITC, Clone: 30-F11, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB40-F-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15951968
  • 225.00 € / 500µg
Estimated Shipment: 26-09-2024
to see stock.
Description
Description
Specifications
Specifications
CD45
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2b κ
Flow Cytometry
30-F11
FITC
NP_001255215
Rat
500 μg
Primary
Store at 2-4°C.