Learn More
CD45.2 Mouse anti-Mouse, CoraLite 647, Clone: 104, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL647-65072-100TESTS
Product Code. 16801116
  • 121.00 € / 100 tests
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
The CD45.2 antibody from Proteintech is a mouse monoclonal antibody generated with b10.s mouse thymocytes and splenocytes of mouse CD45.2. This antibody recognizes mouse antigen. The CD45.2 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifications
Specifications
CD45.2
Monoclonal
5 μL/Test
Mouse
Primary
Mouse
Flow Cytometry
104
CoraLite 647
100 Tests
19264
IgG2a